missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASP9, Mouse, Clone: LAAP6 96 2-22, Abnova™
Mouse monoclonal antibody raised against CASP9.
Brand: Abnova MAB8426.100ug
This item is not returnable.
View return policy
Specifications
| CASP9 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Western Blot | |
| LAAP6 96 2-22 | |
| Mouse monoclonal antibody raised against CASP9. | |
| In PBS (0.08% sodium azide) | |
| A mixture of recombinant human CASP9 and mouse myeloma cells corresponding to CASP9. | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction