Learn More
CARD11, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CARD11.
Brand: Abnova H00084433-A01.50uL
Description
The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein has a domain structure similar to that of CARD14 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. [provided by RefSeq
Sequence: KYFLPYHPPQRRMNLKGIQLQRAKSPISLKRTSDFQAKGHEEEGTDASPSSCGSLPITNSFTKMQPPRSRSSIMSITAEPPGNDSIVRRYKEDAPHRSTVSpecifications
| CARD11 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CARD11. | |
| CARD11 | |
| BIMP3/CARMA1/MGC133069 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_032415 | |
| CARD11 | |
| CARD11 (NP_115791, 481 a.a. to 580 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 84433 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only