missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Canine IgE Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen™ MA516652
This item is not returnable.
View return policy
Description
This antibody recognizes canine IgE, and does not cross react with canine IgM, IgA, IgG1 or IgG2. Western blot analysis against affinity purified IgE shows a single major band of 62-65kDa under reducing conditions.
Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Canine IgE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide | |
| Affinity purified IgE preparation from heavily parasitized and allergic dog serum. | |
| 250 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| E6-2A1 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Canine | |
| Secondary Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction