missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human C-Reactive Protein/CRP Antibody, R&D Systems™

Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
| Antigen | C-Reactive Protein/CRP |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 232007 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P02741 |
| Gene Alias | C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?