Learn More
C-peptide Mouse anti-Human, Clone: C-PEP-01, Abnova™
Mouse monoclonal antibody raised against C-peptide.
Brand: Abnova MAB5025.100ug
Description
After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds. Binding of this mature form of insulin to the insulin receptor (INSR) stimulates glucose uptake. A variety of mutant alleles with changes in the coding region have been identified. There is a read-through gene, INS-IGF2, which overlaps with this gene at the 5' region and with the IGF2 gene at the 3' region. [provided by RefSeq]
Specifications
| C-peptide | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (25 ug/mL) The optimal working dilution should be determined by the end user. | |
| INS | |
| INS | |
| Human C-peptide conjugated with BSA. | |
| RUO | |
| 3630 | |
| Human | |
| Liquid |
| Immunohistochemistry | |
| C-PEP-01 | |
| Mouse monoclonal antibody raised against C-peptide. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| ILPR/IRDN | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts specifically with C-peptide, a part of the Proinsulin molecule. Proinsulin consists of the three parts: C-peptide and two long strands of amino acids (alpha and beta chains; later become linked together to form the Insulin molecule). No cross-reactivity with Insulin or other peptide hormones or proteins was observed. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.