Learn More
BUB1, Mouse, Clone: 2F9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant BUB1.
Brand: Abnova H00000699-M02A.200uL
Description
This gene encodes a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint. Mutations in this gene have been associated with aneuploidy and several forms of cancer. [provided by RefSeq
Sequence: MDTPENVLQMLEAHMQSYKGNDPLGEWERYIQWVEENFPENKEYLITLLEHLMKEFLDKKKYHNDPRFISYCLKFAEYNSDLHQFFEFLYNHGIGTLSSPLYIAWAGHLEAQGELQHASAVLQRGIQNQASpecifications
| BUB1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC028201 | |
| BUB1 | |
| BUB1 (AAH28201,1 a.a. ∽ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 699 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 2F9 | |
| Mouse monoclonal antibody raised against a partial recombinant BUB1. | |
| BUB1 | |
| BUB1A/BUB1L/hBUB1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.