Learn More
BRCA1, Mouse, Clone: RAY, Abnova™
Mouse monoclonal antibody raised against partial recombinant BRCA1.
Brand: Abnova MAB2362.100uL
Description
This gene encodes a nuclear phosphoprotein that plays a role in maintaining genomic stability, and it also acts as a tumor suppressor. The encoded protein combines with other tumor suppressors, DNA damage sensors, and signal transducers to form a large multi-subunit protein complex known as the BRCA1-associated genome surveillance complex (BASC). This gene product associates with RNA polymerase II, and through the C-terminal domain, also interacts with histone deacetylase complexes. This protein thus plays a role in transcription, DNA repair of double-stranded breaks, and recombination. Mutations in this gene are responsible for approximately 40% of inherited breast cancers and more than 80% of inherited breast and ovarian cancers. Alternative splicing plays a role in modulating the subcellular localization and physiological function of this gene. Many alternatively spliced transcript variants, some of which are disease-associated mutations, have been described for this gene, but the full-length natures of only some of these variants has been described. A related pseudogene, which is also located on chromosome 17, has been identified. [provided by RefSeq
Specifications
| BRCA1 | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (1:100) Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| BRCA1 | |
| BRCA1 | |
| Recombinant protein corresponding to amino acids 1314-1864 of human BRCA1. | |
| RUO | |
| 672 | |
| Human | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| RAY | |
| Mouse monoclonal antibody raised against partial recombinant BRCA1. | |
| In PBS (0.09% sodium azide) | |
| BRCAI/BRCC1/IRIS/PSCP/RNF53 | |
| Mouse | |
| 100 μL | |
| Primary | |
| This antibody recognizes the epitope aa 1314-1600. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.