missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Bovine IgA Heavy Chain Secondary Antibody (IL-A71), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68410IR
This item is not returnable.
View return policy
Description
IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Bovine samples. It is validated for ELISA, Immunoprecipitation
Specifications
| IgA Heavy Chain | |
| Monoclonal | |
| DyLight 755 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bovine | |
| Purified |
| ELISA, Immunoprecipitation | |
| IL-A71 | |
| 50mM Sodium Borate | |
| Purified bovine Immunoglobulin | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction