missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Bovine Ig Mix Secondary Antibody
Mouse Monoclonal Secondary Antibody
£413.00 - £828.00
Specifications
| Antigen | Bovine Ig Mix |
|---|---|
| Clone | BIG10-123.1 |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11534990
|
Invitrogen™
SA110639 |
500 μg |
£413.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17860206
|
Invitrogen™
SA5-10360 |
1 mg |
£828.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17890226
|
Invitrogen™
SA5-10361 |
100 μg |
£828.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SA1-10639 detects bovine Immunoglobulin. SA1-10639 has been successfully used in ELISA applications. The SA1-10639 immunogen is purified bovine immunoglobulin.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Bovine Ig Mix | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Bovine | |
| Purified bovine immunoglobulin. | |
| Secondary Antibody |
| BIG10-123.1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Secondary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title