missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMI1, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of BMI1.
Brand: Abnova MAB8718.100ug
This item is not returnable.
View return policy
Specifications
| BMI1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user. | |
| BMI1 | |
| MGC12685/PCGF4/RNF51 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 648 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against synthetic peptide of BMI1. | |
| In PBS, pH 7.4 (0.02% sodium azide, 1% BSA, 50% glycerol) | |
| NM_005180.6 | |
| BMI1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus residues of human BMI1. | |
| 100 μg | |
| Primary | |
| This antibody is specific to BMI1. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction