Learn More
BLNK, Mouse, Clone: 5G9, Abnova™
Mouse monoclonal antibody raised against partial recombinant BLNK.
Brand: Abnova MAB10505.100uL
Description
This gene encodes a cytoplasmic linker or adaptor protein that plays a critical role in B cell development. This protein bridges B cell receptor-associated kinase activation with downstream signaling pathways, thereby affecting various biological functions. The phosphorylation of five tyrosine residues is necessary for this protein to nucleate distinct signaling effectors following B cell receptor activation. Mutations in this gene cause hypoglobulinemia and absent B cells, a disease in which the pro- to pre-B-cell transition is developmentally blocked. Deficiency in this protein has also been shown in some cases of pre-B acute lymphoblastic leukemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| BLNK | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| BLNK | |
| BLNK | |
| Recombinant protein corresponding to human BLNK. | |
| RUO | |
| 29760 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 5G9 | |
| Mouse monoclonal antibody raised against partial recombinant BLNK. | |
| In ascites (0.03% sodium azide) | |
| BASH/BLNK-S/LY57/MGC111051/SLP-65/SLP65 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.