Learn More
BIRC5, Mouse, Clone: 2H5H2, Abnova™
Mouse monoclonal antibody raised against recombinant BIRC5.
Brand: Abnova MAB10414.100uL
Description
This gene is a member of the inhibitor of apoptosis (IAP) gene family, which encode negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but this gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of this gene's expression. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. [provided by RefSeq]
Specifications
| BIRC5 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| BIRC5 | |
| BIRC5 | |
| Recombinant protein corresponding to human BIRC5. | |
| RUO | |
| 332 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| 2H5H2 | |
| Mouse monoclonal antibody raised against recombinant BIRC5. | |
| In ascites (0.03% sodium azide) | |
| API4/EPR-1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.