missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BFSP1, Mouse, Clone: FIL-27, Abnova™
Mouse monoclonal antibody raised against native BFSP1.
Brand: Abnova MAB2423.100ug
This item is not returnable.
View return policy
Specifications
| BFSP1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Frozen sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| BC041483 | |
| BFSP1 | |
| Native from human and bovine lens filament enriched fraction (plasma membrane-cytoskeleton complex). | |
| 100 μg | |
| Primary | |
| Bovine, Human | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| FIL-7B10 | |
| Mouse monoclonal antibody raised against native BFSP1. | |
| Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
| CP115/CP94/FILENSIN/LIFL-H | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 631/281644 | |
| Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction