Learn More
BCL7B, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant BCL7B.
Brand: Abnova H00009275-A01.50uL
Description
The protein encoded by this gene contains a region that is highly similar to the N-terminal segment of BCL7A protein. The BCL7A protein is encoded by the gene known to be directly involved in a three-way gene translocation in a Burkitt lymphoma cell line. This gene is located at a chromosomal region commonly deleted in Williams syndrome. The function of this gene has not yet been determined. [provided by RefSeq
Sequence: AHTSDFRTDDSQPPTLGQEILEEPSLPSSEVADEPPTLTKEEPVPLETQVVEEEEDSGAPPLKRFCVDQPTVPQTASESSpecifications
| BCL7B | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant BCL7B. | |
| BCL7B | |
| BCL7B | |
| BCL7B (NP_001698, 124 a.a. to 202 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9275 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001707 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only