Learn More
BCL2L12, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant BCL2L12.
Brand: Abnova H00083596-A01.50uL
Description
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a Bcl-2 homology domain 2 (BH2). The function of this gene has not yet been determined. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq
Sequence: RSKLVRLSSDSFARLVELFCSRDDSSRPSRACPGPSPPSPEPLARLALAMELSRRVAGLGGTLAGLSVEHVHSFTPWIQAHGGWEGILAVSPVDLNLPLDSpecifications
| BCL2L12 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant BCL2L12. | |
| BCL2L12 | |
| MGC120313/MGC120314/MGC120315 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC007724 | |
| BCL2L12 | |
| BCL2L12 (AAH07724, 174 a.a. to 273 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 83596 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only