missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL10, Mouse, Clone: 4F8E8H8, Abnova™
Mouse monoclonal antibody raised against recombinant BCL10.
Brand: Abnova MAB5567.100ug
This item is not returnable.
View return policy
Specifications
| BCL10 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:2000-1:5000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:500) Immunohistochemistry (Frozen sections) (1:200-1:1000) The optimal working dilution should be determined by the end | |
| BCL10 | |
| BCL10 | |
| Recombinant protein corresponding to BCL10. | |
| RUO | |
| 8915 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| 4F8E8H8 | |
| Mouse monoclonal antibody raised against recombinant BCL10. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| CARMEN/CIPER/CLAP/c-E10/mE10 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction