missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAP31, Mouse, Clone: A1/182, Abnova™
Mouse monoclonal antibody raised against BCAP31.
Brand: Abnova MAB5149.100ug
This item is not returnable.
View return policy
Specifications
| BCAP31 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000) Immunocytochemistry (1:1000) The optimal working dilution should be determined by the end user. | |
| BCAP31 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Recognizes human and monkey BAP31. | |
| Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| A1/182 | |
| Mouse monoclonal antibody raised against BCAP31. | |
| Lyophilized from purified antibody | |
| BCAP31 | |
| Monkey BAP31. | |
| RUO | |
| 696663 | |
| Human, Monkey | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction