Learn More
BATF, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant BATF.
Brand: Abnova H00010538-A01
Description
The protein encoded by this gene is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events. [provided by RefSeq
Sequence: EKNRIAAQKSRQRQTQKADTLHLESEDLEKQNAALRKEIKQLTEELKYFTSVLNSHEPLCSVLAASTPSPPEVVYSAHAFHQPHVSSPRFQPSpecifications
| BATF | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant BATF. | |
| BATF | |
| B-ATF/BATF1/SFA-2/SFA2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006399 | |
| BATF | |
| BATF (NP_006390, 34 a.a. to 125 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 10538 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only