Learn More
B3GAT1, Mouse, Clone: 6-13-19-1, Abnova™
Mouse monoclonal antibody raised against native B3GAT1.
Brand: Abnova MAB5257.1mL
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized. [provided by RefSeq
Specifications
| B3GAT1 | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (1:5) The optimal working dilution should be determined by the end user. | |
| B3GAT1 | |
| B3GAT1 | |
| Native purified B3GAT1 from JURKAT T-cell line. | |
| RUO | |
| 27087 | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry | |
| 6-13-19-1 | |
| Mouse monoclonal antibody raised against native B3GAT1. | |
| In tissue culture supernatant | |
| CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1 | |
| Mouse | |
| 1 mL | |
| Primary | |
| This antibody reacts with a 110 kD antigen present on the peripheral blood lymphocyte subset (primarily large granular lymphocytes) identified by the HNK-1 monoclonal antibody. It recognizes part of the Natural Killer cell population. CD57 is furthermore present on cells in neuroectodermal tissue. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.