Learn More
ATP6V0A1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ATP6V0A1.
Brand: Abnova H00000535-A01.50uL
Description
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene encodes one of three A subunit proteins and the encoded protein is associated with clathrin-coated vesicles. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GDYVHKSVFIIFFQGDQLKNRVKKICEGFRASLYPCPETPQERKEMASGVNTRIDDLQMVLNQTEDHRQRVLQAAAKNIRVWFIKVRSpecifications
| ATP6V0A1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ATP6V0A1. | |
| ATP6V0A1 | |
| ATP6N1/ATP6N1A/DKFZp781J1951/Stv1/VPP1/Vph1/a1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005177 | |
| ATP6V0A1 | |
| ATP6V0A1 (NP_005168, 212 a.a. to 298 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 535 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only