missing translation for 'onlineSavingsMsg'
Learn More

ATP1B1, Mouse, Clone: 464.8 (also known as 8A), Abnova™

Mouse monoclonal antibody raised against native ATP1B1.

Brand:  Abnova MAB2323.100uL

Product Code. 16035576

  • £535.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

ATP1B1
Monoclonal
Unconjugated
Western Blot (1:2500-1:10000) The optimal working dilution should be determined by the end user.
ATP1B1
Mouse
100 μL
Primary
This antibody is specific to Na, K-ATPase beta 1 subunit.
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
IgG2a κ
Western Blot
464.8 (also known as
Mouse monoclonal antibody raised against native ATP1B1.
In buffer containing 0.09% sodium azide
ATP1B1
Native purified rabbit ATP1B1.
RUO
100008749
Bovine, Canine, Human, Pig, Rabbit, Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.