missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATM, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against partial recombinant ATM.
Brand: Abnova MAB6132.100ug
This item is not returnable.
View return policy
Specifications
| ATM | |
| Monoclonal | |
| Mouse monoclonal antibody raised against partial recombinant ATM. | |
| In PBS, pH 7.4 | |
| Recombinant GST fusion protein corresponding to amino acids 2577-3056 of ATM. | |
| RUO | |
| This antibody recognizes full-length ATM, a 370 kD nuclear phosphoprotein, which is involved in the autooal recessive disease ataxia telangiectasia (AT). | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1-10 ug/mL) Immunoprecipitation (1-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction