Learn More
ARL6IP5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ARL6IP5.
Brand: Abnova H00010550-A01.50uL
Description
Expression of this gene is affected by vitamin A. The encoded protein of this gene may be associated with the cytoskeleton. A similar protein in rats may play a role in the regulation of cell differentiation. The rat protein binds and inhibits the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport. [provided by RefSeq
Sequence: MDVNIAPLRAWDDFFPGSDRFARPDFRDISKWNNRVVSNLLYYQTNYLVVAAMMISIVGFLSPFSpecifications
| ARL6IP5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ARL6IP5. | |
| ARL6IP5 | |
| DERP11/GTRAP3-18/HSPC127/JWA/PRAF3/addicsin/hp22/jmx | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006407 | |
| ARL6IP5 | |
| ARL6IP5 (NP_006398, 1 a.a. to 64 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 10550 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only