Learn More
ARL2BP, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ARL2BP protein.
Brand: Abnova H00023568-B01P.50ug
Description
ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2.GTP with high affinity but does not interact with ARL2.GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2.GTP but lack of ARL2 GTPase-activating protein activity. [provided by RefSeq
Sequence: MDALEGESFALSFSSASDAEFDAVVGYLEDIIMDDEFQLLQRNFMDKYYLEFEDTEENKLIYTPIFNEYISLVEKYIEEQLLQRIPEFNMAAFTTTLQHHKDEVAGDIFDMLLTFTDFLAFKEMFLDYRAEKEGRGLDLSSGLVVTSLCKSSSLPASQNNLRHSpecifications
| ARL2BP | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ARL2BP protein. | |
| ARL2BP | |
| BART/BART1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23568 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_012106.3 | |
| ARL2BP | |
| ARL2BP (NP_036238.1,1 a.a. ∽ 163 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only