missing translation for 'onlineSavingsMsg'
Learn More

ARID4A, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant ARID4A.

Brand:  Abnova H00005926-A01.50uL

Product Code. 16124155

  • £209.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a ubiquitously expressed nuclear protein. It binds directly, with several other proteins, to retinoblastoma protein (pRB) which regulates cell proliferation. pRB represses transcription by recruiting the encoded protein. This protein, in turn, serves as a bridging molecule to recruit HDACs and, in addition, provides a second HDAC-independent repression function. The encoded protein possesses transcriptional repression activity. Multiple alternatively spliced transcripts have been observed for this gene, although not all transcript variants have been fully described. [provided by RefSeq

Sequence: AEESQEGLCERESANGFETNVASGTCSIIVQERESREKGQKRPSDGNSGLMAKKQKRTPKRTSAAAKNEKNGTGQSSDSEDLPVLDNSSKCTPVKHLNVSKPQKLAR
Specifications

Specifications

ARID4A
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant ARID4A.
ARID4A
RBBP1/RBP-1/RBP1
Mouse
50 μL
Primary
Human
Antisera
ELISA, Western Blot
Unconjugated
50% glycerol
NM_002892
ARID4A
ARID4A (NP_002883, 1033 a.a. to 1139 a.a) partial recombinant protein with GST tag.
RUO
5926
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only