missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGDIB, Mouse, Clone: 10D774, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of ARHGDIB.
Brand: Abnova MAB0058.100ug
This item is not returnable.
View return policy
Description
Sequence: YVQHTYRTGVKVDSpecifications
| ARHGDIB | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| ARHGDIB | |
| ARHGDIB | |
| A synthetic peptide corresponding to amino acids 125-137 of human ARHGDIB. | |
| RUO | |
| 397 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Western Blot | |
| 10D774 | |
| Mouse monoclonal antibody raised against synthetic peptide of ARHGDIB. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| D4/GDIA2/GDID4/LYGDI/Ly-GDI/RAP1GN1/RhoGDI2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction