Learn More
APOO, Mouse, Clone: 2F1, Abnova™
Mouse monoclonal antibody raised against partial recombinant APOO.
Brand: Abnova MAB10492.100uL
Description
This gene is a member of the apolipoprotein family. Members of this protein family are involved in the transport and metabolism of lipids. The encoded protein associates with HDL, LDL and VLDL lipoproteins and is characterized by chondroitin-sulfate glycosylation. This protein may be involved in preventing lipid accumulation in the myocardium in obese and diabetic patients. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 3, 4, 5, 12 and 16
Specifications
| APOO | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| APOO | |
| APOO | |
| Recombinant protein corresponding to human APOO. | |
| RUO | |
| 79135 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 2F1 | |
| Mouse monoclonal antibody raised against partial recombinant APOO. | |
| In ascites (0.03% sodium azide) | |
| FAM121B/MGC4825/MYO25/My025 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.