Learn More
APOC2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant APOC2.
Brand: Abnova H00000344-A01.50uL
Description
The protein encoded by this gene is secreted in plasma where it is a component of very low density lipoprotein. This protein activates the enzyme lipoprotein lipase, which hydrolyzes triglycerides and thus provides free fatty acids for cells. Mutations in this gene cause hyperlipoproteinemia type IB, characterized by hypertriglyceridemia, xanthomas, and increased risk of pancreatitis and early atherosclerosis. [provided by RefSeq
Sequence: TQQPQQDEMPSPTLLTQVKESLSSYWESAKTAAQNLYEKTYLPAVDEKLRDLYSKSTAAMSTYTGIFTDQVLSVLKGEESpecifications
| APOC2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant APOC2. | |
| APOC2 | |
| MGC75082 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC005348 | |
| APOC2 | |
| APOC2 (AAH05348, 23 a.a. to 101 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 344 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only