Learn More
ANXA11, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ANXA11.
Brand: Abnova H00000311-A01.50uL
Description
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56kDa antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified. [provided by RefSeq
Sequence: IEKSICREMSGDLEEGMLAVVKCLKNTPAFFAERLNKAMRGAGTKDRTLIRIMVSRSETDLLDIRSEYKRMYGKSLYHDISGDTSGDYRKILLKICGGNDSpecifications
| ANXA11 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ANXA11. | |
| ANXA11 | |
| ANX11/CAP50 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001157 | |
| ANXA11 | |
| ANXA11 (NP_001148, 406 a.a. to 505 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 311 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only