missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMY2A/AMY2B Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native AMY2A/AMY2B.
Brand: Abnova MAB2214.100ug
This item is not returnable.
View return policy
Specifications
| AMY2A/AMY2B | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native AMY2A/AMY2B. | |
| In PBS, pH 7.4 | |
| AMY2A | |
| Native purified AMY2A/AMY2B from human pancreas. | |
| RUO | |
| 279/280 | |
| Human | |
| Liquid |
| ELISA, Radioimmune Assays (RIA) | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| AMY2/AMY2B/PA | |
| Mouse | |
| 100 μg | |
| Primary | |
| These antibodies specifically recognize human pancreatic amylase. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction