missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human AMPD2 protein.
Brand: Abnova H00000271-B01P.50ug
This item is not returnable.
View return policy
Description
Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.[supplied by OMIM
Sequence: MASEARGGLGAPPLQSARSLPGPAPCLKHFPLDLRTSMDGKCKEIAEELFTRSLAESELRSAPYEFPEESPIEQLEERRQRLERQISQDVKLEPDILLRAKQDFLKTDSDSDLQLYKEQGEGQGDRSLRERDVLEREFQRVTISGEEKCGVPFTDLLDAAKSVVRALFIREKYMALSLQSFCPTTRRYLQQLAEKPLETRTYEQGPDTPVSADAPVHPPALEQHPYEHCEPSTMPGDLGLGLRMVRGVVHVYTRREPDEHCSEVELPYPDLQEFVADVNVLMALIINGPIKSFCYRRLQYLSSKFQMHVLLNEMKELAAQKKVPHRDFYNIRKVDTHIHASSCMNQKHLLRFIKRAMKRHLEEIVHVEQGREQTLREVFESMNLTAYDLSVDTLDVHADRNTFHRFDKFNAKYNPIGESVLREIFIKTDNRVSGKYFAHIIKEVMSDLEESKYQNAELRLSIYGRSRDEWDKLARWAVMHRVHSPNVRWLVQVPRLFDVYRTKGQLANFQEMLENIFLPLFEATVHPASHPELHLFLEHVDGFDSVDDESKPENHVFNLESPLPEAWVEEDNPPYAYYLYYTFANMAMLNHLRRQRGFHTFVLRPHCGEAGPIHHLVSAFMLAENISHGLLLRKAPVLQYLYYLAQIGIAMSPLSNNSLFLSYHRNPLPEYLSRGLMVSLSTDDPLQFHFTKEPLMEEYSIATQVWKLSSCDMCELARNSVLMSGFSHKVKSHWLGPNYTKEGPEGNDIRRTNVPDIRVGYRYETLCQELALITQAVQSEMLETIPEEAGITMSPGPQSpecifications
| AMPD2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human AMPD2 protein. | |
| AMPD2 | |
| AMPD2 | |
| AMPD2 (NP_631895.1,1 a.a. ∽ 798 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_139156.2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 271 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction