Learn More
ALYREF, Mouse, Clone: 11G5, Abnova™
Mouse monoclonal antibody raised against recombinant ALYREF.
Brand: Abnova MAB2468.100uL
Description
The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins. [provided by RefSeq
Specifications
| ALYREF | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| ALYREF | |
| ALYREF | |
| Recombinant protein corresponding to human ALYREF. | |
| RUO | |
| 10189 | |
| Frog, Human | |
| Liquid |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 11G5 | |
| Mouse monoclonal antibody raised against recombinant ALYREF. | |
| In PBS (0.09% sodium azide) | |
| ALY/ALY/REF/BEF/REF/THOC4 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Detects a band of approximately 32 KDa. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.