Learn More
ALG5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ALG5.
Brand: Abnova H00029880-A01.50uL
Description
This gene encodes a member of the glycosyltransferase 2 family. The encoded protein participates in glucosylation of the oligomannose core in N-linked glycosylation of proteins. The addition of glucose residues to the oligomannose core is necessary to ensure substrate recognition, and therefore, effectual transfer of the oligomannose core to the nascent glycoproteins. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: RDTQCGFKLFTREAASRTFSSLHVERWAFDVELLYIAQFFKIPIAEIAVNWTEIEGSKLVPFWSWLQMGKDLLFIRLRYLTGAWRLEQTRKMNSpecifications
| ALG5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ALG5. | |
| ALG5 | |
| RP11-421P11.2/bA421P11.2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_013338 | |
| ALG5 | |
| ALG5 (NP_037470, 232 a.a. to 324 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 29880 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only