Learn More
ALDH1A1, Mouse, Clone: 5A11, Abnova™
Mouse monoclonal antibody raised against recombinant ALDH1A1.
Brand: Abnova MAB10443.100uL
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme. This gene encodes a cytosolic isoform, which has a high affinity for aldehydes. [provided by RefSeq
Specifications
| ALDH1A1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| ALDH1A1 | |
| ALDH1A1 | |
| Recombinant protein corresponding to human ALDH1A1. | |
| RUO | |
| 216 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 5A11 | |
| Mouse monoclonal antibody raised against recombinant ALDH1A1. | |
| In ascitic (0.03% sodium azide) | |
| ALDC/ALDH-E1/ALDH1/ALDH11/MGC2318/PUMB1/RALDH1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.