Learn More
AKT2, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against partial recombinant protein of AKT2.
Brand: Abnova MAB11138.50ug
Description
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. The gene was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins. [provided by RefSeq
Specifications
| AKT2 | |
| Monoclonal | |
| Mouse monoclonal antibody raised against partial recombinant protein of AKT2. | |
| In PBS, pH 7.4 (1% BSA, 50% glycerol, 0.02% sodium azide) | |
| PKBB/PKBBETA/PRKBB/RAC-BETA | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 208 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Flow Cytometry, Western Blot | |
| Unconjugated | |
| ELISA (1:5000-1:20000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user. | |
| AKT2 | |
| AKT2 | |
| Recombinant protein corresponding to N-terminus of human AKT2. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.