missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ahi1, Mouse, Clone: 645s3, Abnova™
Mouse monoclonal antibody raised against partial recombinant Ahi1.
Brand: Abnova MAB8962.100uL
This item is not returnable.
View return policy
Specifications
| Ahi1 | |
| Monoclonal | |
| 1 ug/uL | |
| Mouse monoclonal antibody raised against partial recombinant Ahi1. | |
| In PBS, pH 7.4 (0.02% sodium azide) | |
| 1700015F03Rik/Ahi-1/D10Bwg0629e | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 52906 | |
| Mouse | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 645s3 | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) Immunoprecipitation (1-2 ug/200-800 ug protein) IHC (1:200-1:1000) The optimal working dilution should be determined. | |
| Ahi1 | |
| Ahi1 | |
| Recombinant protein corresponding to C-terminus of mouse Ahi-1. | |
| 100 μL | |
| Primary | |
| This antibody only specifically reacts with murine Ahi-1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction