Learn More
AGXT, Mouse, Clone: AT2T4, Abnova™
Mouse monoclonal antibody raised against partial recombinant AGXT.
Brand: Abnova MAB8226.100uL
Description
This gene is expressed only in the liver and the encoded protein is localized mostly in the peroxisomes, where it is involved in glyoxylate detoxification. Mutations in this gene, some of which alter subcellular targetting, have been associated with type I primary hyperoxaluria. [provided by RefSeq
Specifications
| AGXT | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| AGXT | |
| AGXT | |
| Recombinant protein corresponding to amino acids 330-392 of human AGXT. | |
| RUO | |
| 189 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
| ELISA, Immunofluorescence | |
| AT2T4 | |
| Mouse monoclonal antibody raised against partial recombinant AGXT. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| AGT/AGT1/AGXT1/PH1/SPAT/SPT/TLH6 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.