Learn More
ACRV1, Mouse, Clone: Hs-14, Abnova™
Mouse monoclonal antibody raised against native purified ACRV1.
Brand: Abnova MAB3824.100ug
Description
The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation. [provided by RefSeq
Specifications
| ACRV1 | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| VCP | |
| VCP | |
| Native purified human VCP. | |
| RUO | |
| 7415 | |
| Human, Mouse | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| Hs-14 | |
| Mouse monoclonal antibody raised against native purified ACRV1. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| IBMPFD/MGC131997/MGC148092/MGC8560/TERA/p97 | |
| Mouse | |
| 100 μg | |
| Primary | |
| The antibody Hs-14 reacts with VCP (valosin-containing protein) a 220 kDa protein previously identified under the general name “intra-acrosomal protein”. | |
| Store at 4°C. Do not freeze. | |
| IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.