Learn More
ACACB, Mouse, Clone: 3E8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ACACB.
Brand: Abnova H00000032-M02A.200uL
Description
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. ACC-beta is thought to control fatty acid oxidation by means of the ability of malonyl-CoA to inhibit carnitine-palmitoyl-CoA transferase I, the rate-limiting step in fatty acid uptake and oxidation by mitochondria. ACC-beta may be involved in the regulation of fatty acid oxidation, rather than fatty acid biosynthesis. There is evidence for the presence of two ACC-beta isoforms. [provided by RefSeq
Sequence: IWGKMTDSKPITKSKSEANLIPSQEPFPASDNSGETPQRNGEGHTLPKTPSQAEPASHKGPKDAGRRRNSLPPSHQKPPRNPLSSSDAAPSPELQANGTSpecifications
| ACACB | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_001093 | |
| ACACB | |
| ACACB (NP_001084,22 a.a. ∽ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 32 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3E8 | |
| Mouse monoclonal antibody raised against a partial recombinant ACACB. | |
| ACACB | |
| ACC2/ACCB/HACC275 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.