missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-like Protein 3/ANGPTL3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF136
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 3/ANGPTL3 Polyclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Angiopoietin-like Protein 3/ANGPTL3 | |
| Polyclonal | |
| Unconjugated | |
| Q9R182 | |
| ANGPTL3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Angiopoietin-like Protein 3/ANGPTL3 Ser17-Thr455 Accession # Q9R182 | |
| 100 μg | |
| Primary | |
| Detects mouse Angiopoietin-like Protein 3/ANGPTL3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-1, rhAngiopoietin-2, recombinant mouse Angiopoietin-3, rhAngiopoietin-4, and rhAngiopoietin-like factor/CDT6 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27329 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction