missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Angiopoietin-like 6 (NP_660136.1) VersaClone cDNA
Specifications
Specifications
| Gene | Angiopoietin-like Protein 6/ANGPTL6 |
| Accession Number | NP_660136.1 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1387 bp |
| Gene ID (Entrez) | 83854 |
| Gene Alias | AGF, AGFAngiopoietin-related protein 5, Angiopoietin like Protein 6, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ANGPTL6, ARP5, ARP5angiopoietin-related protein 6 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?