missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-like 3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB136-SP
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 3/ANGPTL3 Monoclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Mouse samples. It is validated for Western Blot.
Specifications
| Angiopoietin-like Protein 3/ANGPTL3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9R182 | |
| ANGPTL3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Angiopoietin-like 3 Ser17-Thr455 Accession # Q9R182 | |
| 25 μg | |
| Primary | |
| Detects mouse Angiopoietin-like 3 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Angoipoietin-1, rhAngoipoietin-2, rhAngoipoietin-4, rmAngoipoietin-3, and cornea-derived transcript 6 (CDT6), an Angiopoietin-like factor. | |
| Mouse | |
| Purified |
| Western Blot | |
| 128610 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27329 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction