missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mouse Amphiregulin ELISA Kit (Chemiluminescence)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Mouse Amphiregulin ELISA Kit (Chemiluminescence) measures mouse Amphiregulin in serum, plasma and other biological fluids.
Specifikationer
Specifikationer
| Assay Range | 7.81-500 pg/mL |
| Assay Sensitivity | 4.69 pg/mL |
| Cross Reactivity | This kit recognizes Mouse AR in samples. No significant cross-reactivity or interference between Mouse AR and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 374 |
| Gene Symbol | AREG |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering