missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Aminopeptidase PILS/ARTS1 Antibody, R&D Systems™

Goat Polyclonal Antibody
£156.00 - £433.00
Specifications
| Antigen | Aminopeptidase PILS/ARTS1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Neutralization 12 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18788318
![]() |
R&D Systems
AF2500-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18050242
|
R&D Systems
AF2500 |
100 μg |
£433.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase PILS/ARTS1 Polyclonal specifically detects Aminopeptidase PILS/ARTS1 in Mouse samples. It is validated for Western Blot, Neutralization.Specifikationer
| Aminopeptidase PILS/ARTS1 | |
| Western Blot 0.1 ug/mL, Neutralization 12 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| Adipocyte-derived leucine aminopeptidase, A-LAP, A-LAPALAP, Aminopeptidase PILS, ARTS1, ARTS1aminopeptidase regulator of TNFR1 shedding, ARTS-1PILSAP, EC 3.4.11, EC 3.4.11.-, EC 3.4.11.1, endoplasmic reticulum aminopeptidase 1, endoplasmic reticulum aminopeptidase associated with antigen processing, ERAAP1, ERAP1, KIAA0525APPILS, PILS-AP, PILS-APERAAP, Puromycin-insensitive leucyl-specific aminopeptidase, Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator | |
| ERAP1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Q6GTP5 | |
| 51752 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Aminopeptidase PILS/ARTS1 Ser27-Leu930 Accession # Q6GTP5 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Aminopeptidase PILS/ARTS1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human ARTS1 is observed. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel