missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Aminopeptidase PILS/ARTS1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2500-SP
This item is not returnable.
View return policy
Description
Aminopeptidase PILS/ARTS1 Monoclonal specifically detects Aminopeptidase PILS/ARTS1 in Mouse samples. It is validated for Western Blot.
Specifications
| Aminopeptidase PILS/ARTS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_109636 | |
| ERAP1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Aminopeptidase PILS/ARTS1 Ser27-Leu930 Accession # NP_109636 | |
| 25 μg | |
| Primary | |
| Detects mouse Aminopeptidase PILS/ARTS1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human Aminopeptidase PILS/ARTS is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 325908 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adipocyte-derived leucine aminopeptidase, A-LAP, A-LAPALAP, Aminopeptidase PILS, ARTS1, ARTS1aminopeptidase regulator of TNFR1 shedding, ARTS-1PILSAP, EC 3.4.11, EC 3.4.11.-, EC 3.4.11.1, endoplasmic reticulum aminopeptidase 1, endoplasmic reticulum aminopeptidase associated with antigen processing, ERAAP1, ERAP1, KIAA0525APPILS, PILS-AP, PILS-APERAAP, Puromycin-insensitive leucyl-specific aminopeptidase, Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51752 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction