missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Aminopeptidase N/CD13 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 15 publications
Brand: R&D Systems AF2335-SP
This item is not returnable.
View return policy
Description
Aminopeptidase N/CD13 Polyclonal specifically detects Aminopeptidase N/CD13 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF-ready.
Specifications
| Aminopeptidase N/CD13 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 290 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 0.1-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| P97449 | |
| ANPEP | |
| Mouse myeloma cell line NS0-derived recombinant mouse Aminopeptidase N/CD13 Lys69-Ser966 Accession # P97449 | |
| 25 μg | |
| Primary | |
| Detects mouse Aminopeptidase N/CD13 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction