missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse AMICA/JAML Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3106
This item is not returnable.
View return policy
Description
AMICA/JAML Polyclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Western Blot.
Specifications
| AMICA/JAML | |
| Polyclonal | |
| Biotin | |
| Q80UL9 | |
| AMICA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse AMICA/JAML Leu23-Leu281 Accession # Q80UL9 | |
| 50 μg | |
| Primary | |
| Detects mouse AMICA/JAML in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human AMICA is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 120425 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction