missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ALK-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF770-SP
This item is not returnable.
View return policy
Description
ALK-1 Polyclonal specifically detects ALK-1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| ALK-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 94 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q61288 | |
| ACVRL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ALK-1 Asp23-Pro119 Accession # Q61288 | |
| 25 μg | |
| Primary | |
| Detects mouse ALK-1 in ELISAs and Western blots. In sandwich immunoassays, less than 5% cross-reactivity with recombinant human ALK-1 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction