missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ALK-1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB770
Additional Details : Weight : 0.00970kg
Description
ALK-1 Monoclonal specifically detects ALK-1 in Mouse samples. It is validated for Western Blot.Specifications
ALK-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
94 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
143430 | |
Western Blot 1 ug/mL | |
Q61288 | |
ACVRL1 | |
Mouse myeloma cell line NS0-derived recombinant mouse ALK-1 Asp23-Pro119 Accession # Q61288 | |
500 μg | |
Primary | |
Detects mouse ALK-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Activin RIA, rhActivin RIB, or rhALK-1. | |
Mouse | |
Purified |
For Research Use Only