missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse ADAMTS1 ELISA Kit

Product Code. 17134903
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17134903

Brand: Invitrogen™ EM4RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Mouse ADAMTS1 quantitates mouse ADAMTS1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse ADAMTS1.

ADAMTS-1 is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene contains two disintegrin loops and three C-terminal TS motifs and has anti-angiogenic activity. The expression of this gene may be associated with various inflammatory processes as well as development of cancer cachexia. This gene is likely to be necessary for normal growth, fertility, and organ morphology and function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 1; a disintegrin and metalloproteinase with thrombospondin motifs 1 (ADAMTS-1); a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 1; a disintegrin-like and metallopeptidse (reprolysin type) with thrombospondin type 1 motif, 1; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 1; ADAM metallopeptidase with thrombospondin type 1 motif, 1; ADAMs; ADAMTS; ADAM-TS 1; Adamts1; ADAM-TS1; ADAMTS-1; C3-C5; human metalloproteinase with thrombospondin type 1 motifs; KIAA1346; metalloendopeptidases; METH1; METH-1
Assay Range 0.4 to 100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 11504
Gene Symbol ADAMTS1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target ADAMTS1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.